Anti-CSF3R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 38660]

Supplier: R & D Systems
FAB381P
220017-672EA 454.09 USD
220017-672
Anti-CSF3R Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 38660]
Antibodies
Type: Primary
Antigen: CSF3R
Clonality: Monoclonal
Clone: 38660
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR