Anti-MCSF1R Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 460615]

Supplier: R & D Systems
FAB3818A
220017-667EA 561.11 USD
220017-667
Anti-MCSF1R Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 460615]
Antibodies
Type: Primary
Antigen: MCSF1R
Clonality: Monoclonal
Clone: 460615
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR