Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11830]

Supplier: R & D Systems
FAB3791P-025 FAB3791P-100
102882-592EA 241.51 USD
102882-592 102882-590
Anti-CD4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 11830]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 11830
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR