Anti-CD4 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 11830]

Supplier: R & D Systems
FAB3791N-100 FAB3791N-025
102882-586EA 146.06 USD
102882-586 102882-588
Anti-CD4 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 11830]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 11830
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR