Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11830]

Supplier: R & D Systems
FAB3791A-100 FAB3791A-025
102882-570EA 128.71 USD
102882-570 102882-572
Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11830]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 11830
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR