Anti-PLXNB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 559830]

Supplier: R & D Systems
FAB37491A
220017-661EA 594.37 USD
220017-661
Anti-PLXNB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 559830]
Antibodies
Type: Primary
Antigen: PLXNB1
Clonality: Monoclonal
Clone: 559830
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR