Anti-FPR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 350418]

Supplier: R & D Systems
FAB3744F
220017-659EA 446.86 USD
220017-659
Anti-FPR1 Mouse Monoclonal Antibody (Fluorescein) [clone: 350418]
Antibodies
Type: Primary
Antigen Symbol: FPR1
Antigen Name: formyl peptide receptor 1
Antigen Synonyms: FMLP, FPR
Clonality: Monoclonal
Clone: 350418
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR