Anti-FPR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 350418]

Supplier: R & D Systems
FAB3744A
220017-658EA 608.83 USD
220017-658
Anti-FPR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 350418]
Antibodies
Type: Primary
Antigen Symbol: FPR1
Antigen Name: formyl peptide receptor 1
Antigen Synonyms: FMLP, FPR
Clonality: Monoclonal
Clone: 350418
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR