Anti-ADGRE5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 58772]

Supplier: R & D Systems
FAB3734P
102882-566EA 454.09 USD
102882-566
Anti-ADGRE5 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 58772]
Antibodies
Type: Primary
Antigen: ADGRE5
Clonality: Monoclonal
Clone: 58772
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR