Anti-NGFR Mouse Monoclonal Antibody (APC) [clone: 74902]

Supplier: R & D Systems
FAB367A
102882-552EA 608.83 USD
102882-552
Anti-NGFR Mouse Monoclonal Antibody (APC) [clone: 74902]
Antibodies
Type: Primary
Antigen Symbol: NGFR
Antigen Name: nerve growth factor receptor
Antigen Synonyms: CD271, Gp80-LNGFR, TNFRSF16, p75(NTR), p75NTR
Clonality: Monoclonal
Clone: 74902
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR