Anti-CD44 (v6) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2F10]

Supplier: R & D Systems
FAB3660P
220017-651EA 268.98 USD
220017-651
Anti-CD44 (v6) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 2F10]
Antibodies
Type: Primary
Antigen: CD44 (v6)
Clonality: Monoclonal
Clone: 2F10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR