Anti-CD44 (v6) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 2F10]

Supplier: R & D Systems
FAB3660G
102882-350EA 404.92 USD
102882-350
Anti-CD44 (v6) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 2F10]
Antibodies
Type: Primary
Antigen: CD44 (v6)
Clonality: Monoclonal
Clone: 2F10
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR