Anti-CD44 (v6) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2F10]

Supplier: R & D Systems
FAB3660A
220017-649EA 517.72 USD
220017-649
Anti-CD44 (v6) Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 2F10]
Antibodies
Type: Primary
Antigen: CD44 (v6)
Clonality: Monoclonal
Clone: 2F10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR