Anti-AGTR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 36485]

Supplier: R & D Systems
FAB3659P
220017-648EA 513.38 USD
220017-648
Anti-AGTR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 36485]
Antibodies
Type: Primary
Antigen: AGTR2
Clonality: Monoclonal
Clone: 36485
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR