Anti-C5AR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 347214]

Supplier: R & D Systems
FAB3648P
220017-643EA 543.75 USD
220017-643
Anti-C5AR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 347214]
Antibodies
Type: Primary
Antigen: C5AR1
Clonality: Monoclonal
Clone: 347214
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR