Anti-CD48 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 394607]

Supplier: R & D Systems
FAB3644P
220017-640EA 480.12 USD
220017-640
Anti-CD48 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 394607]
Antibodies
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 394607
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR