Anti-CMKLR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 84939]

Supplier: R & D Systems
FAB362A
220017-637EA 577.02 USD
220017-637
Anti-CMKLR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 84939]
Antibodies
Type: Primary
Antigen: CMKLR1
Clonality: Monoclonal
Clone: 84939
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR