Anti-ITGAL Mouse Monoclonal Antibody (Fluorescein) [clone: 345913]

Supplier: R & D Systems
FAB3595F
220017-634EA 438.18 USD
220017-634
Anti-ITGAL Mouse Monoclonal Antibody (Fluorescein) [clone: 345913]
Antibodies
Type: Primary
Antigen: ITGAL
Clonality: Monoclonal
Clone: 345913
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR