Anti-MET Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 95106]

Supplier: R & D Systems
FAB3582P
220017-632EA 582.8 USD
220017-632
Anti-MET Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 95106]
Antibodies
Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: 95106
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR