Anti-MET Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 95106]

Supplier: R & D Systems
FAB3582A
220017-630EA 614.62 USD
220017-630
Anti-MET Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 95106]
Antibodies
Type: Primary
Antigen: MET
Clonality: Monoclonal
Clone: 95106
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR