Anti-ITGA7 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 334908]

Supplier: R & D Systems
FAB3518A
220017-618EA 614.62 USD
220017-618
Anti-ITGA7 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 334908]
Antibodies
Type: Primary
Antigen: ITGA7
Clonality: Monoclonal
Clone: 334908
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR