Anti-CLEC2D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 402659]

Supplier: R & D Systems
FAB3480A
220017-610EA 614.62 USD
220017-610
Anti-CLEC2D Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 402659]
Antibodies
Type: Primary
Antigen: CLEC2D
Clonality: Monoclonal
Clone: 402659
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR