Anti-FPR2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 304405]

Supplier: R & D Systems
FAB3479N
102882-518EA 543.75 USD
102882-518
Anti-FPR2 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 304405]
Antibodies
Type: Primary
Antigen Symbol: FPR2
Antigen Name: formyl peptide receptor 2
Antigen Synonyms: ALXR, FMLP-R-II, FMLPX, FPR2A, FPRH1, FPRH2, FPRL1, HM63, LXA4R
Clonality: Monoclonal
Clone: 304405
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR