Anti-CCR10 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 314305]

Supplier: R & D Systems
FAB3478G-025 FAB3478G-100
102882-510EA 608.83 USD
102882-510 102882-508
Anti-CCR10 Rat Monoclonal Antibody (Alexa Fluor® 488) [clone: 314305]
Antibodies
Type: Primary
Antigen: CCR10
Clonality: Monoclonal
Clone: 314305
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR