Anti-CCR10 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 314305]

Supplier: R & D Systems
FAB3478A-025 FAB3478A-100
102882-500EA 253.08 USD
102882-500 102882-502
Anti-CCR10 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 314305]
Antibodies
Type: Primary
Antigen: CCR10
Clonality: Monoclonal
Clone: 314305
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR