Anti-CCR7 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4B12]

Supplier: R & D Systems
FAB3477P-025 FAB3477P-100
102882-496EA 177.88 USD
102882-496 102882-498
Anti-CCR7 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 4B12]
Antibodies
Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 4B12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR