Anti-CD200R1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 380525]

Supplier: R & D Systems
FAB3414P
220017-594EA 480.12 USD
220017-594
Anti-CD200R1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 380525]
Antibodies
Type: Primary
Antigen: CD200R1
Clonality: Monoclonal
Clone: 380525
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR