Anti-CD200R1 Mouse Monoclonal Antibody (Fluorescein) [clone: 380525]

Supplier: R & D Systems
FAB3414F
220017-593EA 389.02 USD
220017-593
Anti-CD200R1 Mouse Monoclonal Antibody (Fluorescein) [clone: 380525]
Antibodies
Type: Primary
Antigen: CD200R1
Clonality: Monoclonal
Clone: 380525
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR