Anti-CD200R1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 380525]

Supplier: R & D Systems
FAB3414A
220017-592EA 543.75 USD
220017-592
Anti-CD200R1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 380525]
Antibodies
Type: Primary
Antigen: CD200R1
Clonality: Monoclonal
Clone: 380525
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR