Anti-TNFRSF4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 443318]

Supplier: R & D Systems
FAB3388A
220017-586EA 600.15 USD
220017-586
Anti-TNFRSF4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 443318]
Antibodies
Type: Primary
Antigen: TNFRSF4
Clonality: Monoclonal
Clone: 443318
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR