Anti-CD101 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 307707]

Supplier: R & D Systems
FAB3368A
220017-583EA 513.38 USD
220017-583
Anti-CD101 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 307707]
Antibodies
Type: Primary
Antigen: CD101
Clonality: Monoclonal
Clone: 307707
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR