Anti-BTLA Goat Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))

Supplier: R & D Systems
FAB3354C
102882-484EA 577.02 USD
102882-484
Anti-BTLA Goat Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
Antibodies
Type: Primary
Antigen: BTLA
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR