Anti-PTGDR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 301109]

Supplier: R & D Systems
FAB33381P
102882-476EA 513.38 USD
102882-476
Anti-PTGDR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 301109]
Antibodies
Type: Primary
Antigen: PTGDR2
Clonality: Monoclonal
Clone: 301109
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR