Anti-LAG3 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 631501]

Supplier: R & D Systems
FAB33281N
102882-466EA 594.37 USD
102882-466
Anti-LAG3 Rat Monoclonal Antibody (Alexa Fluor® 700) [clone: 631501]
Antibodies
Type: Primary
Antigen: LAG3
Clonality: Monoclonal
Clone: 631501
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR