Anti-CXCR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 48311]

Supplier: R & D Systems
FAB331P-100 FAB331P-025
102882-268EA 177.88 USD
102882-268 102882-270
Anti-CXCR2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 48311]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 48311
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR