Anti-CXCR2 Mouse Monoclonal Antibody (Fluorescein) [clone: 48311]

Supplier: R & D Systems
FAB331F-025 FAB331F-100
102882-262EA 373.11 USD
102882-262 102882-260
Anti-CXCR2 Mouse Monoclonal Antibody (Fluorescein) [clone: 48311]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 48311
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR