Anti-CXCR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 48311]

Supplier: R & D Systems
FAB331A-025 FAB331A-100
102882-252EA 160.52 USD
102882-252 102882-254
Anti-CXCR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 48311]
Antibodies
Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 48311
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR