Anti-CXCR1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 42705]

Supplier: R & D Systems
FAB330N-100 FAB330N-025
102882-460EA 160.52 USD
102882-460 102882-462
Anti-CXCR1 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 42705]
Antibodies
Type: Primary
Antigen: CXCR1
Clonality: Monoclonal
Clone: 42705
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR