Anti-MSLN Rat Monoclonal Antibody (Fluorescein) [clone: 420411]

Supplier: R & D Systems
FAB32652F
220017-560EA 420.83 USD
220017-560
Anti-MSLN Rat Monoclonal Antibody (Fluorescein) [clone: 420411]
Antibodies
Type: Primary
Antigen: MSLN
Clonality: Monoclonal
Clone: 420411
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR