Anti-MSLN Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 420411]

Supplier: R & D Systems
FAB32652A
220017-559EA 639.2 USD
220017-559
Anti-MSLN Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 420411]
Antibodies
Type: Primary
Antigen: MSLN
Clonality: Monoclonal
Clone: 420411
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR