Anti-CDH13 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 392411]

Supplier: R & D Systems
FAB3264P
102882-440EA 513.38 USD
102882-440
Anti-CDH13 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 392411]
Antibodies
Type: Primary
Antigen: CDH13
Clonality: Monoclonal
Clone: 392411
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR