Anti-FLT1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 49560]

Supplier: R & D Systems
FAB321A
220017-554EA 582.8 USD
220017-554
Anti-FLT1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 49560]
Antibodies
Type: Primary
Antigen Symbol: FLT1
Antigen Name: fms-related tyrosine kinase 1
Antigen Synonyms: FLT, FLT-1, VEGFR-1, VEGFR1
Clonality: Monoclonal
Clone: 49560
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR