Anti-BSG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: TRA-1-85]

Supplier: R & D Systems
FAB3195P
220017-553EA 513.38 USD
220017-553
Anti-BSG Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: TRA-1-85]
Antibodies
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: TRA-1-85
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR