Anti-BSG Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: TRA-1-85]

Supplier: R & D Systems
FAB3195N
102882-434EA 519.17 USD
102882-434
Anti-BSG Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: TRA-1-85]
Antibodies
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: TRA-1-85
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR