Anti-BSG Mouse Monoclonal Antibody (Fluorescein) [clone: TRA-1-85]

Supplier: R & D Systems
FAB3195F
220017-552EA 438.18 USD
220017-552
Anti-BSG Mouse Monoclonal Antibody (Fluorescein) [clone: TRA-1-85]
Antibodies
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: TRA-1-85
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR