Anti-BSG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: TRA-1-85]

Supplier: R & D Systems
FAB3195A
220017-551EA 594.37 USD
220017-551
Anti-BSG Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: TRA-1-85]
Antibodies
Type: Primary
Antigen: BSG
Clonality: Monoclonal
Clone: TRA-1-85
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR