Anti-TEK Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 83715]

Supplier: R & D Systems
FAB3131N
102882-424EA 519.17 USD
102882-424
Anti-TEK Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 83715]
Antibodies
Type: Primary
Antigen: TEK
Clonality: Monoclonal
Clone: 83715
Conjugation: Alexa Fluor® 700
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR