Anti-LINGO1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 332237]

Supplier: R & D Systems
FAB30861A
102882-412EA 594.37 USD
102882-412
Anti-LINGO1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 332237]
Antibodies
Type: Primary
Antigen: LINGO1
Clonality: Monoclonal
Clone: 332237
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR