Anti-LILRA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 586326]

Supplier: R & D Systems
FAB30851A
220017-545EA 594.37 USD
220017-545
Anti-LILRA1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 586326]
Antibodies
Type: Primary
Antigen: LILRA1
Clonality: Monoclonal
Clone: 586326
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR