Anti-GPA33 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 402104]

Supplier: R & D Systems
FAB3080P
102882-410EA 533.63 USD
102882-410
Anti-GPA33 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 402104]
Antibodies
Type: Primary
Antigen Symbol: GPA33
Antigen Name: glycoprotein A33 (transmembrane)
Antigen Synonyms: A33
Clonality: Monoclonal
Clone: 402104
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR